Labelling tapes
Supplier: SEMADENI
0162 4339 4340 4335 0353
SEMA0162EA 0 SGD
SEMA0162 SEMA4339 SEMA4340 SEMA4335 SEMA0353
Labelling tapes
Tapes
Labeling tape for glass, plastic, porcelain or metal.
  • Tear resistant
  • Can be easily written on
  • Adhesive strength at low temperature
Length: 66,0 m
Different colours available.
Caution: Do not autoclave.