Tubing connectors, L- shaped
Supplier: SEMADENI
0545
SEMA0545EA 0 SGD
SEMA0545
Tubing connectors, L- shaped
Fittings and Connectors
Made of PP.
  • Very good chemical resistance
  • Very good temperature resistance