Spray bottles
Supplier: SEMADENI
1055
SEMA1055EA 0 SGD
SEMA1055
Spray bottles
Bottles Spray Bottles
Robust and handy LDPE spray bottles with stable construction.
  • Emitted volume by pressure 1,3 ml
  • Neck Ø 25 mm
  • Good chemical resistance even against aggressive media
The spray head and nozzle are made of PP and PE. By rotation, the nozzle can be closed or adjusted to produce a very fine vapour through to a powerful jet.
Order Now
SPECIFICATIONS
Environmentally Preferable