Containers, rectangular
Supplier: SEMADENI
1294
SEMA1294EA 0 SGD
SEMA1294
Containers, rectangular
Sample Containers General Purpose Containers
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.