Storage and Transport Boxes
Supplier: SEMADENI
1308
SEMA1308EA 41.5 SGD
SEMA1308
Storage and Transport Boxes
Boxes Storage Boxes
Boxes, made of PP, have a solid design and are stackable.
  • Standardized on pallet mass
  • With reinforcing ribs on the side
  • Containers with a height of 235 mm or more are not suitable for liquids
  • Covers for mounting without the device for locking or snapping
Ordering information: Lids have to be ordered separately.