Tubing, PVC
Supplier: SEMADENI
1340
SEMA1340EA 0 SGD
SEMA1340
Tubing, PVC
Tubing Transfer Tubing
Made of PVC, clear.
  • Flexible
  • Can be used from −20 to + 60 °C
  • Not suitable for vacuum or printing
Order Now
SPECIFICATIONS
Environmentally Preferable