Centrifuge tubes
Supplier: SEMADENI
1567
SEMA1567EA 0 SGD
SEMA1567
Centrifuge tubes
Tubes Centrifuge Tubes
PP, clear.
  • Stable
  • Autoclavable
  • Temperature resistant up to 130 °C
  • Excellent chemical resistance
Round bottom with rim.