Jars with overlapping lid
Supplier: SEMADENI
1713 3390 2960 2961 2112 4684 4685
SEMA1713EA 0 SGD
SEMA1713 SEMA3390 SEMA2960 SEMA2961 SEMA2112 SEMA4684 SEMA4685
Jars with overlapping lid
Jars General Use Jars
Jars, made of PS, clear, with closing lid.
  • Good chemical resistance
  • Easy to clean
Suitable for ointments, powders, and granules.