Transport and Storage Container
Supplier: SEMADENI
1940
SEMA1940EA 0 SGD
SEMA1940
Transport and Storage Container
Boxes Transport Boxes
Transport and storage containers, made of PP, grey, with reinforced rim and two handles. They are not suitable for liquids.