Disposal canisters, with screw cap
Supplier: SEMADENI
2013
SEMA2013EA 0 SGD
SEMA2013
Disposal canisters, with screw cap
Bottles Jerricans
HDPE, coloured
  • Good chemical resistance
  • Good temperature resistance
  • With a hollow handle, filling opening and vent screw
  • Stackable
For collecting chemicals.