Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Supplier: SEMADENI
2025 2024
SEMA2025EA 0 SGD
SEMA2025 SEMA2024
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Bottles Plastic Bottles
HDPE, translucent, with thread.
  • Good chemical resistance
  • Good temperature resistance
Ordering information: Screw caps must be ordered separately.