Draining racks, wall mounted
Supplier: SEMADENI
2110
SEMA2110EA 0 SGD
SEMA2110
Draining racks, wall mounted
Racks Draining Racks
White, PVC drain channel, polyethylene coated metal rack.
  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning
Order Now
SPECIFICATIONS
Environmentally Preferable