Magnetic stirring bars, cylindrical
Supplier: SEMADENI
2209 0244 0257 0258 0259 0320
SEMA2209EA 0 SGD
SEMA2209 SEMA0244 SEMA0257 SEMA0258 SEMA0259 SEMA0320
Magnetic stirring bars, cylindrical
Stirrers Magnetic Stirring Bars
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
  • Chemical resistance
  • Smooth surface and round ends
  • Temperature resistance upto 275 °C
Order Now
SPECIFICATIONS
Environmentally Preferable