Disposable cuvettes
Supplier: SEMADENI
2531
SEMA2531EA 0 SGD
SEMA2531
Disposable cuvettes
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes for photometric measurements in the visible range (350 to 800 nm).