Disposable syringes, 3-piece
Supplier: SEMADENI
2955
SEMA2955EA 0 SGD
SEMA2955
Disposable syringes, 3-piece
Syringes Syringes, Medical
The piston and its seal are made of silicone rubber.
  • Available with centred and eccentric tip
Certifications: Conforms with ISO 7886, 2955, 5993, 10521.
Order Now
SPECIFICATIONS
Environmentally Preferable