Spatulas, disposable
Supplier: SEMADENI
3339
SEMA3339EA 52.3 SGD
SEMA3339
Spatulas, disposable
Spatulas
PS, white. These disposable spatulas are ideal for handling creams, paints or other viscous media.
  • Durable, slightly elastic
  • Available in different sizes and designs