Pasteur pipettes, disposable, LDPE
Supplier: SEMADENI
3352
SEMA3352EA 130 SGD
SEMA3352
Pasteur pipettes, disposable, LDPE
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable, non-sterile.
  • Almost unbreakable transfer pipettes
  • Good chemical resistance and can be used at low temperatures
  • Pipettes are made from one piece: No risk of leakage
  • Dustproof packaged in convenient standard boxes
Order Now
SPECIFICATIONS
Environmentally Preferable