Jars, with slip lid
Supplier: SEMADENI
3998 1997 3395
SEMA3998EA 0 SGD
SEMA3998 SEMA1997 SEMA3395
Jars, with slip lid
Jars General Use Jars
PS, clear
  • Good chemical resistance
  • Temperature resistance from −10 to 70 °C
Ideal for sampling and storage of solid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904