Laboratory trays
Supplier: SEMADENI
4065
SEMA4065EA 0 SGD
SEMA4065
Laboratory trays
Trays/Dishes/Pots Laboratory Trays/Dishes
Trays made of white co-polyester.
  • Stable
  • Temperature resistant from −40 to +220 °C
  • Suitable for use as a sterilisation or microwave tray
With rounded corners and smooth surfaces.