Jars with tamper-evident screw cap
Supplier: SEMADENI
4285 4287
SEMA4285EA 0 SGD
SEMA4285 SEMA4287
Jars with tamper-evident screw cap
Jars General Use Jars
Jars,made of PP, with HDPE screw caps.
  • Easy to use
  • Can be easily opened and closed
  • Suitable for food storage
Order Now
SPECIFICATIONS
Environmentally Preferable