Bottles, narrow neck, round or square, HDPE, with screw caps
Supplier: SEMADENI
4291
SEMA4291EA 0 SGD
SEMA4291
Bottles, narrow neck, round or square, HDPE, with screw caps
Bottles Plastic Bottles
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Un approved 3H1/X250 for fluids
With tamper evident screw cap.