Bottles, narrow neck, octagonal, PET, with screw caps
Supplier: SEMADENI
4858 4634
SEMA4858EA 0 SGD
SEMA4858 SEMA4634
Bottles, narrow neck, octagonal, PET, with screw caps
Bottles Plastic Bottles
PET, clear.
  • Sterile
  • Good chemical and temperature resistance
  • With tamper evident screw cap
Perfect for sample collection, storage and transport.