Tubing connectors, Y-shaped, PP
Supplier: SEMADENI
4660
SEMA4660EA 5.7 SGD
SEMA4660
Tubing connectors, Y-shaped, PP
Fittings and Connectors
Connectors, Y- shaped, made of PP, natural color.
  • Strong design with long, conical connections for large tolerance ranges
  • Temperature resistant up to 120 °C, sterilisable
Order Now
SPECIFICATIONS
Environmentally Preferable