Sharps disposal boxes, Multi-Safe quick
Supplier: SEMADENI
4779
SEMA4779EA 3.2 SGD
SEMA4779
Sharps disposal boxes, Multi-Safe quick
Bins Sharps Containers
The practical safety box for user-friendly disposal of sharp, pointed and dangerous objects for both stationary and ambulatory use.
  • Round container form
  • Openings of different sizes
  • Containers can be fastened to a number of different holders to prevent them from overturning
  • Closure cap does not spring back. This means nothing gets in the way during removal and stripping
  • Various detaching devices