Safety baskets
Supplier: SEMADENI
4954 2699 4953
SEMA4954EA 0 SGD
SEMA4954 SEMA2699 SEMA4953
Safety baskets
Baskets Safety Transporting Baskets
PP, coloured.
  • Anti-static
  • Space-saving, stackable
  • Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.
Order Now
SPECIFICATIONS
Environmentally Preferable