Jars, wide mouth, with screw cap
Supplier: SEMADENI
5099
SEMA5099EA 0 SGD
SEMA5099
Jars, wide mouth, with screw cap
Jars General Use Jars
Jars, made of HDPE, white colour. Suitable for used as a packaging container.
Ordering information: Lids available in different colours must be ordered separately.