Catheter syringes
Supplier: SEMADENI
6188
SEMA6188EA 0 SGD
SEMA6188
Catheter syringes
Syringes Syringes, Medical
These catheter syringes have a PP barrel with silicone rubber piston.
  • Can be uesd as suction and irrigation syringes
  • Also used in technical purposes for pastes, glues and more
These syringes are gas sterilisable.
Packaging: Individually packed.