Sample cups
Supplier: SEMADENI
6203
SEMA6203EA 0 SGD
SEMA6203
Sample cups
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, translucent, with HDPE screw cap.
  • Good chemical resistance
  • Good temperature resistance
  • With graduation up to 100 ml
  • Tightly sealing HDPE screw top (not autoclavable)
  • Large labelling area
Ideal for hygienic and odorless urine sample handling.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904