Bag openers for Stomacher® bags
Supplier: Seward (FermionX)
BA6095
SEWABA6095EA 0 SGD
SEWABA6095
Bag openers for Stomacher® bags
Racks Bag Racks
Made from high grade plastic polymer.
  • Compatible with Stomacher® 80 bags
  • Stable on bench-top or scales
  • Easily cleaned with most lab disinfectants
These bag openers hold open bags for easy addition of sample without risk of instrument contact on the bag.