Racks for Stomacher® bags
Supplier: Seward (FermionX)
BA6096
SEWABA6096EA 327 SGD
SEWABA6096
Racks for Stomacher® bags
Racks Bag Racks
Made from high grade plastic polymer.
  • Perfect for loading closure bags preenrichments into the incubator
  • Variety of capacities for each Stomacher® machine
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Bag racks provide the perfect storage for Stomacher® bag sample handling, helping to provide a clean and productive working environment.