Syringes, Cavro
Supplier: Trajan Scientific and Medical
008140
SGEA008140EA 0 SGD
SGEA008140
Syringes, Cavro
Syringes Syringes for Chromatography
These cavro syringes are a type of instrument syringes used in chromatography.
  • Luer lock termination
  • Gas tight plunger with CAVRO XL mounting hole