GC capillary column tools
Supplier: Trajan Scientific and Medical
0625011
SGEA0625011EA 0 SGD
SGEA0625011
GC capillary column tools
Cutters
The capillary cutting tool is used for burr free cutting of fused silica tubing.
  • Reduces likelihood of blockages
  • Ensures square ends for 'Press fit' style connectors
  • Clean entry for on-column injection