Pipette stands, hanging
Supplier: IBI SCIENTIFIC
PST-51
SHETPST-51EA 0 SGD
SHETPST-51
Pipette stands, hanging
Pipette Stands
These four-place pipette holders have an additional place for a mechanical pipette filler. The pipette rack is constructed of durable cast acrylic for years of reliable use, and the fluorescent orange color will add colour to the lab. These are orientented such that they can be placed on the right or left side of the fume hood.
  • Holds four standard pipettes and a media filler pipette
  • Attaches to hanging surface with powerful double-sided tape
The PST-50 is oriented so that the pipette filler will be located on the right side, while the pipette fillerMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable