Coating thickness gauge (Fe/NFe)
Supplier: Industrial Physics
LD0800
SIMELD0800EA 0 SGD
SIMELD0800
Coating thickness gauge (Fe/NFe)
Coating Testers
This coating thickness gauge uses automatic magnetic and eddy current principles.
  • Automatic power off
  • F for magnetic induction and N for eddy currents probes
  • Zero point calibration and multi-point calibration (up to four points)
  • Possible to recall and delete specified readings or delete group readings
  • High and low limits alarm, for each group
It can measure the thickness of painted objects, or corrosion protective layers on both ferrous (iron or steel) and non-ferrous (aluminium, brass, copper and non-magnetic steel) metals.
Delivery information: More Product Information