Hardness test sets
Supplier: Industrial Physics
SP0010 SP0015
SIMESP0010EA 0 SGD
SIMESP0010 SIMESP0015
Hardness test sets
Analysers Texture Analysers
These hardness tests are pocket instruments for testing the hardness and wear/scratch resistance of materials such as coatings, lacquers, plastics or related products.
  • Body in anodised aluminium, tips in tungsten carbide
  • Can be used on both flat and curved surface
  • Clearly visible laser-engraved scale on the blue background
  • Ergonomic design with anti-slip texture on holding point
  • Available in two versions (Standard or Pro version)
Tip is drawn over the surface with a defined constant pressure. The pressure on the tip can be changed using the slide orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable