Tripod stand for flow cups
Supplier: Industrial Physics
VF2061
SIMEVF2061EA 0 SGD
SIMEVF2061
Tripod stand for flow cups
Tripods
The spirit level is easy to balance thanks to the adjustable feet of the unit.
  • Stainless steel ring and PVC legs
  • Tripod stand with built-in spirit level
Suitable for all types of viscosity cups.