Temperature control jackets for viscosity cups
Supplier: Industrial Physics
VF2067 VF2068
SIMEVF2067EA 0 SGD
SIMEVF2067 SIMEVF2068
Temperature control jackets for viscosity cups
Controllers Temperature Controllers
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
  • Adjustable feet for easy leveling
  • Built-in spirit level to set jacket exactly horizontal
  • Glass plate to prevent dripping before measurement
  • Elbow-type quick release couplings
  • Titanium anodised aluminium
Delivery information: Supplied with jacket with built-in spirit level, glass plate and tube connection.
Order Now
SPECIFICATIONS
Environmentally Preferable