Urine collection container, Urisafe®
Supplier: Simport Scientific
B350
SIMPB350EA 0 SGD
SIMPB350
Urine collection container, Urisafe®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
A ³/₄ L capacity polypropylene and polyethylene container anatomically designed with a large opening for ease of patient use. A large handle and special spout assists pouring and reduces the risk of aerosol contamination. Graduated in 100 ml increments and 50 ml increments for more accurate reading with the container placed on end for 24 hour urine collection. Large 79 mm opening.
  • Unique snap valve pouring spout
  • Leakproof with controlled, safer dripless pouring
  • Resistant to hydrochloric acid, freeze/thawing and EtO sterilisation