Specimen bottle, B352
Supplier: Simport Scientific
B352 B352-1
SIMPB352EA 0 SGD
SIMPB352 SIMPB352-1
Specimen bottle, B352
Bottles Drosophila Bottles
Polypropylene bottle useful as a Drosophila stock bottle.
  • Graduated from 0 to 170 ml and 0 to 6 oz.
  • Can also be used as a drosophila stock bottle
  • Snap cap supplied separately
  • Autoclavable
Bottles feature a dip-stick well for small volume testing and a pour spout for dripless pouring. The square base makes this bottle very stable.
Ordering information: Snap cap must be purchased separately.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable