Beakers, triangular, Tricorn™
Supplier: Simport Scientific
B700-100 B700-1L B700-250 B700-400 B700-800 B700-50
SIMPB700-100EA 102 SGD
SIMPB700-100 SIMPB700-1L SIMPB700-250 SIMPB700-400 SIMPB700-800 SIMPB700-50
Beakers, triangular, Tricorn™
Beakers
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Packaging: All sizes are available 100 per case.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904