Sample cups, Fibrin
Supplier: Simport Scientific
B720-40
SIMPB720-40EA 56.3 SGD
SIMPB720-40
Sample cups, Fibrin
Sample Containers Sample Cups for Analysers
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Precision molded for constant volume and uniform heat transfer.