Urine containers, SpecTainer™ II
Supplier: Simport Scientific
C567-60Y
SIMPC567-60YEA 0 SGD
SIMPC567-60Y
Urine containers, SpecTainer™ II
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP urine container with leak resistant PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Graduated, except 60 ml container
  • Ridges around base offer a better grip during opening and closing
  • Stackable, shatter resistant
  • Designed with a straight side format
  • Moulded ridges around lid make it easy to open and close
Warning label has space for patient identification. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345).
Order Now
SPECIFICATIONS
Environmentally Preferable