Petri dishes, square with grid
Supplier: Simport Scientific
D210-16
SIMPD210-16EA 751 SGD
SIMPD210-16
Petri dishes, square with grid
Culture Dishes Petri Dishes
PS, sterile. Free from optical distortion, these dishes are ideal for phage typing, susceptibility testing, plate counts, and probe assays. Each 13 mm grid is marked numerically in one direction and alphabetically in the other. This dish has four venting ribs into the underside of the lid to prevent condensation build-up.