Inoculating loops and needles, Ino-Loop™
Supplier: Simport Scientific
L200-1 L200-1A L200-2 L200-2A L200-3 L200-3A L200-11 L200-21 L200-31
SIMPL200-1EA 0 SGD
SIMPL200-1 SIMPL200-1A SIMPL200-2 SIMPL200-2A SIMPL200-3 SIMPL200-3A SIMPL200-11 SIMPL200-21 SIMPL200-31
Inoculating loops and needles, Ino-Loop™
Spreaders Inoculating Loops and Needles
PS, sterile. Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface.
  • Cross contamination due to improper sterilisation is eliminated
  • Can be used under hoods without danger
  • Colour-coded for ease of size identification
  • Certificate of conformity available upon request
Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamperproof, zip-lock resealable bags.
Disposable inoculating loops and needles do not require flaming and thus eliminate the risk ofMore Product Information