Bacterial cell spreaders
Supplier: Simport Scientific
L300
SIMPL300EA 187 SGD
SIMPL300
Bacterial cell spreaders
Spreaders Cell Spreaders
PS, sterile.
  • No flame sterilisation needed
  • Designed for easy spreading of cells onto the surface of an agar plate
  • Supplied sterile in individual packs
Handle length: 176 mm
Head spreader width: 36 mm