Disposable base moulds
Supplier: Simport Scientific
M475-1 M475-3 M475-4 M475-5
SIMPM475-1EA 409 SGD
SIMPM475-1 SIMPM475-3 SIMPM475-4 SIMPM475-5
Disposable base moulds
Moulds Moulds for Histology
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
  • Excellent thermal exchange
  • Smooth interior
  • Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Delivery information: Each case contains two dispenser boxes of 500 base moulds.