Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Supplier: Simport Scientific
M480-10T M480-11T M480-12T M480-2T M480-3T M480-4T M480-5T M480-6T M480-7T M480-8T M480-9T M480-10SL M480-11SL M480-12SL M480-2SL M480-3SL M480-4SL M480-5SL M480-6SL M480-7SL M480-8SL M480-9SL
SIMPM480-10TEA 0 SGD
SIMPM480-10T SIMPM480-11T SIMPM480-12T SIMPM480-2T SIMPM480-3T SIMPM480-4T SIMPM480-5T SIMPM480-6T SIMPM480-7T SIMPM480-8T SIMPM480-9T SIMPM480-10SL SIMPM480-11SL SIMPM480-12SL SIMPM480-2SL SIMPM480-3SL SIMPM480-4SL SIMPM480-5SL SIMPM480-6SL SIMPM480-7SL SIMPM480-8SL SIMPM480-9SL
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
  • The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
  • Keep specimens safely submerged
  • Totally resistant to the chemical action of solvents used in histology laboratories
Order Now
SPECIFICATIONS
Environmentally Preferable