E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Supplier: Simport Scientific
M486-6SL M486-5SL M485-10SL M485-11SL M485-12SL M485-2SL M485-3SL M485-4SL M485-6SL M486-10SL M486-11SL M486-12SL M486-2SL M486-3SL M486-4SL M485-5SL M483-11BA M483-6LI M483-11LI M483-12BA M483-12LI M483-2BA M483-2LI M483-3BA M483-3LI M483-4BA M483-4LI M483-5BA M483-6BA M483-7BA M483-7LI M483-8BA M483-8LI M483-9BA M483-9LI M483-5LI M482-6LI M482-11LI M482-12BA M482-12LI M482-2BA M482-2LI M482-3BA M482-3LI M482-4BA M482-4LI M482-5BA M482-6BA M482-10BA M482-7BA M482-7LI M482-8BA M482-8LI M482-9BA M482-9LI M483-10BA M483-10LI M482-5LI M482-11BA M482-10LI
SIMPM486-6SLEA 0 SGD
SIMPM486-6SL SIMPM486-5SL SIMPM485-10SL SIMPM485-11SL SIMPM485-12SL SIMPM485-2SL SIMPM485-3SL SIMPM485-4SL SIMPM485-6SL SIMPM486-10SL SIMPM486-11SL SIMPM486-12SL SIMPM486-2SL SIMPM486-3SL SIMPM486-4SL SIMPM485-5SL SIMPM483-11BA SIMPM483-6LI SIMPM483-11LI SIMPM483-12BA SIMPM483-12LI SIMPM483-2BA SIMPM483-2LI SIMPM483-3BA SIMPM483-3LI SIMPM483-4BA SIMPM483-4LI SIMPM483-5BA SIMPM483-6BA SIMPM483-7BA SIMPM483-7LI SIMPM483-8BA SIMPM483-8LI SIMPM483-9BA SIMPM483-9LI SIMPM483-5LI SIMPM482-6LI SIMPM482-11LI SIMPM482-12BA SIMPM482-12LI SIMPM482-2BA SIMPM482-2LI SIMPM482-3BA SIMPM482-3LI SIMPM482-4BA SIMPM482-4LI SIMPM482-5BA SIMPM482-6BA SIMPM482-10BA SIMPM482-7BA SIMPM482-7LI SIMPM482-8BA SIMPM482-8LI SIMPM482-9BA SIMPM482-9LI SIMPM483-10BA SIMPM483-10LI SIMPM482-5LI SIMPM482-11BA SIMPM482-10LI
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Embedding Cassettes Cassettes for Labeling Instruments
Acetal polymer. E-Z Load™ Histosette® II cassettes are designed for loading Micro Writer® and Shur/Mark® cassette labelling instruments in one simple operation. Supplied in stacks of 50 cassettes, no manual insertion, one cassette at a time, is required. Just load the stack in the magazine, cut and remove the holding tie and place in the labelling instrument ready for printing. Moulded from a special high density polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slotsMore Product Information