Tissue and biopsy cassettes in QuickLoad™Sleeves, Histosette® II
Supplier: Simport Scientific
M486-8SL M486-9SL M485-7SL M485-15SL M485-16SL M485-17SL M485-18SL M486-7SL M485-8SL M485-9SL
SIMPM486-8SLEA 0 SGD
SIMPM486-8SL SIMPM486-9SL SIMPM485-7SL SIMPM485-15SL SIMPM485-16SL SIMPM485-17SL SIMPM485-18SL SIMPM486-7SL SIMPM485-8SL SIMPM485-9SL
Tissue and biopsy cassettes in QuickLoad™Sleeves, Histosette® II
Embedding Cassettes Tissue Processing Cassettes
Made of acetal polymer. Suitable for hoppers accepting plastic sleeves, these cassettes will load in cassette labeling instruments in one simple operation. Save time and money with these convenient stacks of 75 unique cassettes. No more manual insertion, one cassette at a time.
  • Keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Biopsy model designed to hold small biopsy samples securely during the embedding
More Product Information