Tissue processing/embedding cassettes, Histosette® I, M490
Supplier: Simport Scientific
M490-10 M490-12 M490-2 M490-3 M490-4 M490-5 M490-6 M490-7 M490-9 M490-11 M490-8
SIMPM490-10EA 0 SGD
SIMPM490-10 SIMPM490-12 SIMPM490-2 SIMPM490-3 SIMPM490-4 SIMPM490-5 SIMPM490-6 SIMPM490-7 SIMPM490-9 SIMPM490-11 SIMPM490-8
Tissue processing/embedding cassettes, Histosette® I, M490
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer.
  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid eliminates the need for separate steel lids
  • 30° writing angle